Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHRM5
Entrez Gene ID
1133
Gene Name
cholinergic receptor muscarinic 5
Gene Aliases
HM5
Chromosome Location
Chr.15: 33968314 - 34065094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320917.1 NP_001307846.1 2 - 2 2933 112
NM_012125.3 NP_036257.1 3 - 3 2691 112
GenBank mRNA BC041805.1 - 2 - 2 2335 112
BC068528.1 - 2 - 2 2329 112

View Full Product Details