The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000191.2 | NP_000182.2 | 5 - 6 | 543 | 105 |
GenBank mRNA | AK122801.1 | - | 6 - 7 | 1096 | 105 |
AK313869.1 | - | 5 - 6 | 512 | 105 | |
BC010570.1 | - | 5 - 6 | 509 | 105 | |
BT009792.1 | - | 5 - 6 | 499 | 105 | |
CR456884.1 | - | 5 - 6 | 499 | 105 | |
L07033.1 | - | 5 - 6 | 513 | 105 |