Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FMO9P
- Entrez Gene ID
-
116123
- Gene Name
- flavin containing monooxygenase 9 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 166603916 - 166625238 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002925.2 |
|
4 - 5 |
500 |
65 |
GenBank mRNA |
BC014341.1 |
- |
4 - 5 |
504 |
65 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CV028819.1 |
- |
1 - 2 |
171 |
|
Chr. 1 - 166620817 - 166623791 [+] on Build GRCh38 |
65 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
family with sequence similarity 78 member B |
FAM78B |
|
-454.0kb
|
Anti-sense |
Chr. - 166055918 - 166166755 on Build GRCh38 |
Downstream |
pogo transposable element with KRAB domain |
POGK |
BASS2, KRBOX2, LST003 |
217.4kb
|
Sense |
Chr. - 166839404 - 166856344 on Build GRCh38 |
View Full Product Details