The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033085.2 | NP_149076.1 | 3 - 4 | 429 | 60 |
GenBank mRNA | AF249872.1 | - | 3 - 4 | 429 | 60 |
AY101182.1 | - | 3 - 4 | 457 | 60 | |
BC022064.2 | - | 3 - 4 | 430 | 60 |