The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014691.2 | NP_055506.1 | 31 - 32 | 3953 | 144 |
GenBank mRNA | AB011132.3 | - | 31 - 32 | 3881 | 144 |
AK291615.1 | - | 31 - 32 | 3805 | 144 | |
AK315815.1 | - | 31 - 32 | 3883 | 144 | |
BC036913.2 | - | 7 - 8 | 1092 | 144 | |
BC108262.1 | - | 5 - 6 | 709 | 144 | |
BC127111.1 | - | 31 - 32 | 3824 | 144 | |
BC127112.1 | - | 31 - 32 | 3824 | 144 | |
BX647820.1 | - | 30 - 31 | 4597 | 144 | |
EF553519.1 | - | 31 - 32 | 3914 | 144 |