The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022451.10 | NP_071896.8 | 6 - 7 | 829 | 102 |
XM_017016544.1 | XP_016872033.1 | 4 - 5 | 882 | 102 | |
GenBank mRNA | AB077992.1 | - | 6 - 7 | 716 | 102 |
AK091246.1 | - | 6 - 7 | 786 | 102 | |
AK292520.1 | - | 6 - 7 | 811 | 102 | |
BC049850.1 | - | 6 - 7 | 779 | 102 |