The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006028.4 | NP_006019.1 | 6 - 7 | 826 | 82 |
XM_011543063.1 | XP_011541365.1 | 5 - 6 | 939 | 82 | |
XM_017018552.1 | XP_016874041.1 | 5 - 6 | 530 | 82 | |
GenBank mRNA | AF080582.1 | - | 6 - 7 | 755 | 82 |
AF169255.1 | - | 6 - 7 | 829 | 82 | |
AK314268.1 | - | 6 - 7 | 826 | 82 | |
AM293589.1 | - | 5 - 6 | 698 | 82 |