Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXN4
Entrez Gene ID
121643
Gene Name
forkhead box N4
Gene Aliases
-
Chromosome Location
Chr.12: 109277978 - 109309406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213596.2 NP_998761.2 9 - 10 1403 88
XM_011537922.2 XP_011536224.1 9 - 10 1592 88
XM_017018818.1 XP_016874307.1 7 - 8 2378 88
GenBank mRNA AF425596.1 - 7 - 8 1178 88
AF425597.1 - 6 - 7 1053 88
AK131519.1 - 5 - 6 1010 88
BC113073.1 - 2 - 3 403 88
BC146825.1 - 9 - 10 1404 88

View Full Product Details