The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022363.2 | NP_071758.1 | 1 - 2 | 753 | 62 |
GenBank mRNA | AF291181.1 | - | 1 - 2 | 753 | 62 |
BC109230.2 | - | 1 - 2 | 223 | 62 |