The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267576.1 | NP_001254505.1 | 4 - 5 | 932 | 65 |
NM_153210.4 | NP_694942.3 | 4 - 5 | 932 | 65 | |
XM_011523640.1 | XP_011521942.1 | 3 - 4 | 652 | 65 | |
XM_011523641.1 | XP_011521943.1 | 4 - 5 | 377 | 65 | |
XM_011523642.2 | XP_011521944.1 | 4 - 5 | 977 | 65 | |
XM_017024159.1 | XP_016879648.1 | 3 - 4 | 873 | 65 | |
XM_017024160.1 | XP_016879649.1 | 2 - 3 | 859 | 65 | |
XM_017024162.1 | XP_016879651.1 | 4 - 5 | 977 | 65 | |
GenBank mRNA | AJ583817.2 | - | 4 - 5 | 839 | 65 |
AK055188.1 | - | 4 - 5 | 413 | 65 | |
BC136368.1 | - | 4 - 5 | 932 | 65 | |
BC144041.1 | - | 4 - 5 | 932 | 65 | |
BC171759.1 | - | 4 - 5 | 932 | 65 | |
BX648526.1 | - | 2 - 3 | 195 | 65 |