The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138421.2 | NP_612430.2 | 3 - 4 | 378 | 90 |
GenBank mRNA | AB489136.1 | - | 3 - 4 | 328 | 90 |
AK123457.1 | - | 3 - 4 | 372 | 90 | |
BC012010.1 | - | 3 - 4 | 360 | 90 | |
DR155913.1 | - | 3 - 4 | 378 | 90 |