Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAAL1
Entrez Gene ID
113174
Gene Name
serum amyloid A like 1
Gene Aliases
SPACIA1
Chromosome Location
Chr.11: 18080343 - 18106091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138421.2 NP_612430.2 5 - 6 518 86
GenBank mRNA AB489136.1 - 5 - 6 468 86
AK123457.1 - 5 - 6 512 86
BC012010.1 - 5 - 6 500 86
BU150462.1 - 1 - 2 89 86
DR155913.1 - 5 - 6 518 86

View Full Product Details