The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145049.3 | NP_659486.2 | 9 - 10 | 1120 | 128 |
GenBank mRNA | AK057996.1 | - | 9 - 10 | 911 | 128 |
AY444562.1 | - | 9 - 10 | 932 | 128 | |
BC013425.2 | - | 9 - 10 | 932 | 128 |