The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019024.2 | NP_061897.1 | 20 - 21 | 3152 | 93 |
XM_006712034.2 | XP_006712097.1 | 20 - 21 | 3091 | 93 | |
XM_006712035.3 | XP_006712098.1 | 20 - 21 | 3155 | 93 | |
XM_011532934.2 | XP_011531236.1 | 20 - 21 | 3619 | 93 | |
XM_011532935.2 | XP_011531237.1 | 9 - 10 | 1291 | 93 | |
XM_017004378.1 | XP_016859867.1 | 20 - 21 | 3091 | 93 | |
XM_017004379.1 | XP_016859868.1 | 21 - 22 | 3529 | 93 | |
XM_017004380.1 | XP_016859869.1 | 21 - 22 | 3302 | 93 | |
GenBank mRNA | AB037835.1 | - | 11 - 12 | 1599 | 93 |
AK096637.1 | - | 20 - 21 | 3091 | 93 | |
BC150605.1 | - | 20 - 21 | 3137 | 93 | |
BX538008.1 | - | 14 - 15 | 2074 | 93 | |
BX648805.1 | - | 6 - 7 | 1312 | 93 |