The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019024.2 | NP_061897.1 | 21 - 22 | 3315 | 81 |
XM_006712034.2 | XP_006712097.1 | 21 - 22 | 3254 | 81 | |
XM_006712035.3 | XP_006712098.1 | 21 - 22 | 3318 | 81 | |
XM_011532934.2 | XP_011531236.1 | 21 - 22 | 3782 | 81 | |
XM_011532935.2 | XP_011531237.1 | 10 - 11 | 1454 | 81 | |
XM_017004378.1 | XP_016859867.1 | 21 - 22 | 3254 | 81 | |
XM_017004379.1 | XP_016859868.1 | 22 - 23 | 3692 | 81 | |
XM_017004380.1 | XP_016859869.1 | 22 - 23 | 3465 | 81 | |
GenBank mRNA | AB037835.1 | - | 12 - 13 | 1762 | 81 |
AK096637.1 | - | 21 - 22 | 3254 | 81 | |
BC150605.1 | - | 21 - 22 | 3300 | 81 | |
BX538008.1 | - | 15 - 16 | 2237 | 81 | |
BX648805.1 | - | 7 - 8 | 1475 | 81 |