The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019024.2 | NP_061897.1 | 23 - 24 | 3693 | 142 |
XM_006712034.2 | XP_006712097.1 | 23 - 24 | 3632 | 142 | |
XM_006712035.3 | XP_006712098.1 | 23 - 24 | 3696 | 142 | |
XM_011532934.2 | XP_011531236.1 | 23 - 24 | 4160 | 142 | |
XM_011532935.2 | XP_011531237.1 | 12 - 13 | 1832 | 142 | |
XM_017004378.1 | XP_016859867.1 | 23 - 24 | 3632 | 142 | |
XM_017004379.1 | XP_016859868.1 | 24 - 25 | 4070 | 142 | |
XM_017004380.1 | XP_016859869.1 | 24 - 25 | 3843 | 142 | |
GenBank mRNA | AB037835.1 | - | 14 - 15 | 2140 | 142 |
AK096637.1 | - | 23 - 24 | 3632 | 142 | |
BC150605.1 | - | 23 - 24 | 3678 | 142 | |
BX538008.1 | - | 17 - 18 | 2616 | 142 | |
BX648805.1 | - | 9 - 10 | 1853 | 142 |