The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287580.1 | NP_001274509.1 | 7 - 8 | 1774 | 73 |
NM_022128.2 | NP_071411.1 | 6 - 7 | 655 | 73 | |
GenBank mRNA | AJ404857.1 | - | 6 - 7 | 615 | 73 |
AK300989.1 | - | 6 - 7 | 609 | 73 | |
AY643715.1 | - | 6 - 7 | 647 | 73 | |
BC017425.1 | - | 6 - 7 | 657 | 73 | |
BX648909.1 | - | 7 - 8 | 1745 | 73 |