The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006545.4 | NP_006536.3 | 9 - 10 | 1330 | 100 |
XM_005264808.4 | XP_005264865.1 | 9 - 10 | 942 | 100 | |
XM_011533288.2 | XP_011531590.1 | 8 - 9 | 1012 | 100 | |
XM_017005555.1 | XP_016861044.1 | 9 - 10 | 1045 | 100 | |
XM_017005556.1 | XP_016861045.1 | 8 - 9 | 1026 | 100 | |
GenBank mRNA | AF040707.1 | - | 9 - 10 | 990 | 100 |
AK292196.1 | - | 9 - 10 | 989 | 100 | |
AL833426.1 | - | 7 - 8 | 1681 | 100 | |
BC021984.1 | - | 9 - 10 | 1076 | 100 | |
BC056861.1 | - | 9 - 10 | 1028 | 100 |