The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000180.3 | NP_000171.1 | 6 - 7 | 1643 | 72 |
XM_011523816.1 | XP_011522118.1 | 5 - 6 | 1569 | 72 | |
GenBank mRNA | M92432.1 | - | 6 - 7 | 1642 | 72 |