The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007.4 | NP_000998.1 | 1 - 2 | 98 | 140 |
GenBank mRNA | AK311745.1 | - | 1 - 2 | 88 | 140 |
BU192706.1 | - | 1 - 2 | 140 | 140 |