Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS29
Entrez Gene ID
6235
Gene Name
ribosomal protein S29
Gene Aliases
DBA13, S29
Chromosome Location
Chr.14: 49576672 - 49586416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030001.2 NP_001025172.1 1 - 2 132 132
NM_001032.4 NP_001023.1 1 - 2 132 132
GenBank mRNA BU942244.1 - 1 - 2 135 132

View Full Product Details