The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037329.3 | NP_001032406.1 | 4 - 5 | 1026 | 165 |
XM_017017217.1 | XP_016872706.1 | 4 - 5 | 1026 | 165 | |
XM_017017218.1 | XP_016872707.1 | 4 - 5 | 933 | 165 | |
XM_017017219.1 | XP_016872708.1 | 4 - 5 | 934 | 165 | |
GenBank mRNA | AK122736.1 | - | 4 - 5 | 1018 | 165 |
BC040277.1 | - | 4 - 5 | 965 | 165 | |
BC106935.2 | - | 2 - 3 | 165 | ||
BC106936.2 | - | 2 - 3 | 165 |