The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099334.2 | NP_001092804.1 | 2 - 3 | 214 | 135 |
XM_017004074.1 | XP_016859563.1 | 2 - 3 | 227 | 135 | |
XM_017004076.1 | XP_016859565.1 | 2 - 3 | 202 | 135 | |
GenBank mRNA | BC035737.1 | - | 2 - 3 | 214 | 135 |