Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf80
Entrez Gene ID
389073
Gene Name
chromosome 2 open reading frame 80
Gene Aliases
GONDA1
Chromosome Location
Chr.2: 208165347 - 208190258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099334.2 NP_001092804.1 6 - 7 543 165
XM_017004075.1 XP_016859564.1 6 - 7 491 165
XM_017004076.1 XP_016859565.1 6 - 7 531 165
GenBank mRNA BC035737.1 - 6 - 7 543 165

View Full Product Details