The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182577.2 | NP_872383.1 | 3 - 4 | 582 | 126 |
XM_005259545.4 | XP_005259602.1 | 3 - 4 | 755 | 126 | |
XM_017026671.1 | XP_016882160.1 | 2 - 3 | 152 | 126 | |
GenBank mRNA | AK097378.1 | - | 3 - 4 | 582 | 126 |
BC104468.1 | - | 2 - 3 | 371 | 126 | |
BC104469.1 | - | 3 - 4 | 475 | 126 |