The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012486.2 | NP_036618.2 | 10 - 11 | 1398 | 132 |
XM_017001836.1 | XP_016857325.1 | 9 - 10 | 1211 | 132 | |
GenBank mRNA | AK292299.1 | - | 11 - 12 | 1350 | 132 |
U34349.1 | - | 8 - 9 | 1062 | 132 |