The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199692.1 | NP_001186621.1 | 5 - 6 | 1621 | 169 |
NM_001199693.1 | NP_001186622.1 | 4 - 5 | 647 | 169 | |
NM_001199694.1 | NP_001186623.1 | 4 - 5 | 1068 | 169 | |
NM_003040.3 | NP_003031.3 | 5 - 6 | 837 | 169 | |
XM_006716094.3 | XP_006716157.1 | 5 - 6 | 837 | 169 | |
GenBank mRNA | AB209158.1 | - | 5 - 6 | 1621 | 169 |
AK295767.1 | - | 4 - 5 | 647 | 169 | |
AK312699.1 | - | 5 - 6 | 816 | 169 | |
BC004893.2 | - | 5 - 6 | 474 | 169 | |
BC009386.2 | - | 4 - 5 | 1068 | 169 | |
BC009434.1 | - | 4 - 5 | 1068 | 169 | |
BC010069.2 | - | 5 - 6 | 474 | 169 | |
BC028601.2 | - | 4 - 5 | 669 | 169 | |
U62531.1 | - | 5 - 6 | 755 | 169 | |
X62137.1 | - | 5 - 6 | 660 | 169 |