Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FDPS
Entrez Gene ID
2224
Gene Name
farnesyl diphosphate synthase
Gene Aliases
FPPS, FPS, POROK9
Chromosome Location
Chr.1: 155308748 - 155320666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002004.3 NP_001995.1 1 - 2 220 97

View Full Product Details