Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC80
Entrez Gene ID
285175
Gene Name
unc-80 homolog, NALCN activator
Gene Aliases
C2orf21, UNC-80
Chromosome Location
Chr.2: 209771925 - 209999300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032504.1 NP_115893.1 28 - 29 4663 134
NM_182587.3 NP_872393.3 28 - 29 4648 134
XM_005246476.1 XP_005246533.1 29 - 30 4861 134
XM_011511004.1 XP_011509306.1 29 - 30 4903 134
XM_011511005.1 XP_011509307.1 29 - 30 4903 134
XM_011511006.1 XP_011509308.1 29 - 30 4897 134
XM_011511007.1 XP_011509309.1 29 - 30 4867 134
XM_011511008.1 XP_011509310.1 29 - 30 4903 134
XM_011511010.2 XP_011509312.1 29 - 30 4903 134
XM_011511012.1 XP_011509314.1 29 - 30 4903 134
XM_017003884.1 XP_016859373.1 29 - 30 4888 134
XM_017003885.1 XP_016859374.1 29 - 30 4861 134
XM_017003886.1 XP_016859375.1 29 - 30 4903 134
XM_017003887.1 XP_016859376.1 28 - 29 4699 134
XM_017003888.1 XP_016859377.1 28 - 29 4657 134
XM_017003889.1 XP_016859378.1 28 - 29 4657 134
XM_017003890.1 XP_016859379.1 28 - 29 4642 134
XM_017003891.1 XP_016859380.1 28 - 29 4922 134
XM_017003892.1 XP_016859381.1 16 - 17 3706 134
XM_017003893.1 XP_016859382.1 29 - 30 4903 134
XM_017003894.1 XP_016859383.1 7 - 8 1134 134
GenBank mRNA AB058746.2 - 14 - 15 2108 134

View Full Product Details