Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIS18BP1
Entrez Gene ID
55320
Gene Name
MIS18 binding protein 1
Gene Aliases
C14orf106, HSA242977, KNL2, M18BP1
Chromosome Location
Chr.14: 45203190 - 45253402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018353.4 NP_060823.3 5 - 6 1677 123
XM_005267833.4 XP_005267890.1 6 - 7 1724 123
XM_011536923.1 XP_011535225.1 2 - 3 162 123
XM_017021425.1 XP_016876914.1 7 - 8 1705 123
GenBank mRNA AB067490.1 - 5 - 6 1678 123
BC051885.1 - 5 - 6 1454 123
BC065544.1 - 5 - 6 1468 123

View Full Product Details