The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173988.1 | NP_001167459.1 | 12 - 13 | 2035 | 117 |
NM_024718.4 | NP_078994.3 | 12 - 13 | 2032 | 117 | |
GenBank mRNA | AK000963.1 | - | 9 - 10 | 1438 | 117 |
AK001184.1 | - | 11 - 12 | 1619 | 117 | |
AK027586.1 | - | 9 - 10 | 1453 | 117 | |
AK094382.1 | - | 12 - 13 | 2263 | 117 | |
AK098086.1 | - | 4 - 5 | 3123 | 117 | |
AK128154.2 | - | 12 - 13 | 2296 | 117 | |
AK307431.1 | - | 13 - 14 | 1886 | 117 | |
AL713707.1 | - | 5 - 6 | 1008 | 117 | |
BC018960.2 | - | 12 - 13 | 1978 | 117 | |
BC021095.1 | - | 12 - 13 | 1736 | 117 | |
BC024964.2 | - | 3 - 4 | 518 | 117 | |
BC035786.1 | - | 12 - 13 | 1990 | 117 | |
DQ141240.1 | - | 12 - 13 | 1757 | 117 | |
EF156752.1 | - | 12 - 13 | 1759 | 117 |