The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167738.1 | NP_001161210.1 | 11 - 12 | 2340 | 122 |
NM_020443.4 | NP_065176.3 | 13 - 14 | 3671 | 122 | |
XM_006711609.2 | XP_006711672.2 | 11 - 12 | 2306 | 122 | |
XM_006711610.2 | XP_006711673.2 | 10 - 11 | 2282 | 122 | |
XM_006711611.1 | XP_006711674.1 | 11 - 12 | 2422 | 122 | |
XM_011510097.1 | XP_011508399.1 | 13 - 14 | 3350 | 122 | |
XM_011510098.1 | XP_011508400.1 | 12 - 13 | 3326 | 122 | |
XM_011510099.1 | XP_011508401.1 | 12 - 13 | 3326 | 122 | |
XM_011510100.1 | XP_011508402.1 | 12 - 13 | 3179 | 122 | |
XM_011510101.1 | XP_011508403.1 | 11 - 12 | 3155 | 122 | |
XM_011510102.1 | XP_011508404.1 | 11 - 12 | 3155 | 122 | |
XM_017002751.1 | XP_016858240.1 | 12 - 13 | 3179 | 122 | |
XM_017002752.1 | XP_016858241.1 | 10 - 11 | 2282 | 122 | |
XM_017002753.1 | XP_016858242.1 | 12 - 13 | 3326 | 122 | |
GenBank mRNA | AB032977.3 | - | 13 - 14 | 3004 | 122 |
AB033039.1 | - | 7 - 8 | 1183 | 122 | |
AJ488101.1 | - | 13 - 14 | 3202 | 122 | |
AK056158.1 | - | 10 - 11 | 2393 | 122 | |
AY043013.1 | - | 13 - 14 | 3671 | 122 | |
BX648460.1 | - | 11 - 12 | 2307 | 122 |