Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS11F
Entrez Gene ID
389208
Gene Name
transmembrane protease, serine 11F
Gene Aliases
-
Chromosome Location
Chr.4: 68053198 - 68129869 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207407.2 NP_997290.2 9 - 10 1202 132

View Full Product Details