Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCN6
Entrez Gene ID
158062
Gene Name
lipocalin 6
Gene Aliases
LCN5, UNQ643, hLcn5
Chromosome Location
Chr.9: 136744017 - 136748528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198946.2 NP_945184.1 3 - 4 347 114
GenBank mRNA AF303084.1 - 3 - 4 344 114
AY301269.1 - 3 - 4 302 114
AY358598.1 - 3 - 4 327 114
BC062746.1 - 3 - 4 348 114

View Full Product Details