Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRE4P
Entrez Gene ID
326342
Gene Name
adhesion G protein-coupled receptor E4, pseudogene
Gene Aliases
EMR4, EMR4P, FIRE, GPR127, PGR16
Chromosome Location
Chr.19: 6952500 - 6990846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024075.1 10 - 11 1240 149
GenBank mRNA AF489700.1 - 10 - 11 1240 149
AY181245.1 - 6 - 7 749 149

View Full Product Details