The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022048.3 | NP_071331.2 | 4 - 5 | 771 | 144 |
GenBank mRNA | AB042562.1 | - | 3 - 4 | 419 | 144 |
AB042563.1 | - | 3 - 4 | 403 | 144 | |
AF223357.2 | - | 3 - 4 | 489 | 144 | |
AK225960.1 | - | 4 - 5 | 701 | 144 | |
AL832551.1 | - | 4 - 5 | 549 | 144 | |
BC017236.2 | - | 3 - 4 | 220 | 144 | |
BC130613.1 | - | 4 - 5 | 544 | 144 | |
BC136504.1 | - | 4 - 5 | 544 | 144 |