The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022048.3 | NP_071331.2 | 7 - 8 | 1241 | 122 |
GenBank mRNA | AB042562.1 | - | 6 - 7 | 889 | 122 |
AB042563.1 | - | 6 - 7 | 873 | 122 | |
AF223357.2 | - | 6 - 7 | 959 | 122 | |
AK026957.1 | - | 3 - 4 | 348 | 122 | |
AK225960.1 | - | 7 - 8 | 1171 | 122 | |
AL832551.1 | - | 7 - 8 | 1019 | 122 | |
BC017236.2 | - | 6 - 7 | 690 | 122 | |
BC130613.1 | - | 7 - 8 | 1014 | 122 | |
BC136504.1 | - | 7 - 8 | 1014 | 122 |