Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGLEC14
Entrez Gene ID
100049587
Gene Name
sialic acid binding Ig like lectin 14
Gene Aliases
-
Chromosome Location
Chr.19: 51639478 - 51646879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098612.1 NP_001092082.1 5 - 6 1129 143
GenBank mRNA AK298284.1 - 5 - 6 1051 143
AY854038.1 - 5 - 6 1050 143

View Full Product Details