Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17orf99
Entrez Gene ID
100141515
Gene Name
chromosome 17 open reading frame 99
Gene Aliases
UNQ464
Chromosome Location
Chr.17: 78146035 - 78166283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163075.1 NP_001156547.1 2 - 3 121 168
XM_017023998.1 XP_016879487.1 2 - 3 196 168
GenBank mRNA AY358510.1 - 2 - 3 128 168

View Full Product Details