Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf72
Entrez Gene ID
100130988
Gene Name
chromosome 7 open reading frame 72
Gene Aliases
-
Chromosome Location
Chr.7: 50095861 - 50159256 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161834.2 NP_001155306.2 6 - 7 977 158
GenBank mRNA BX092263.1 - 6 - 7 528 158

View Full Product Details