Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
Chromosome Location
Chr.5: 140786136 - 141012344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031410.2 NP_113598.1 1 - 1 2682 85
GenBank mRNA BF089693.1 - 1 - 1 359 85

View Full Product Details