The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030632.2 | NP_085135.1 | 3 - 4 | 306 | 148 |
XM_005258356.1 | XP_005258413.1 | 4 - 5 | 279 | 148 | |
XM_011526205.2 | XP_011524507.1 | 4 - 5 | 7069 | 148 | |
XM_011526206.2 | XP_011524508.1 | 4 - 5 | 3834 | 148 | |
XM_011526209.1 | XP_011524511.1 | 5 - 6 | 463 | 148 | |
XM_011526210.1 | XP_011524512.1 | 5 - 6 | 520 | 148 | |
XM_011526212.1 | XP_011524514.1 | 4 - 5 | 424 | 148 | |
XM_011526213.1 | XP_011524515.1 | 4 - 5 | 517 | 148 | |
XM_017026012.1 | XP_016881501.1 | 4 - 5 | 3681 | 148 | |
XM_017026013.1 | XP_016881502.1 | 5 - 6 | 4043 | 148 | |
XM_017026014.1 | XP_016881503.1 | 5 - 6 | 480 | 148 | |
GenBank mRNA | BE145544.1 | - | 3 - 4 | 306 | 148 |
DB090653.1 | - | 4 - 5 | 280 | 148 |