The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030632.2 | NP_085135.1 | 5 - 6 | 535 | 134 |
XM_005258356.1 | XP_005258413.1 | 6 - 7 | 508 | 134 | |
XM_011526205.2 | XP_011524507.1 | 6 - 7 | 7298 | 134 | |
XM_011526206.2 | XP_011524508.1 | 6 - 7 | 4063 | 134 | |
XM_011526209.1 | XP_011524511.1 | 7 - 8 | 692 | 134 | |
XM_011526210.1 | XP_011524512.1 | 7 - 8 | 749 | 134 | |
XM_011526212.1 | XP_011524514.1 | 6 - 7 | 653 | 134 | |
XM_011526213.1 | XP_011524515.1 | 6 - 7 | 746 | 134 | |
XM_017026012.1 | XP_016881501.1 | 6 - 7 | 3910 | 134 | |
XM_017026013.1 | XP_016881502.1 | 7 - 8 | 4272 | 134 | |
XM_017026014.1 | XP_016881503.1 | 7 - 8 | 709 | 134 | |
GenBank mRNA | AK131454.1 | - | 6 - 7 | 500 | 134 |