The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030632.2 | NP_085135.1 | 7 - 8 | 772 | 147 |
XM_005258356.1 | XP_005258413.1 | 8 - 9 | 745 | 147 | |
XM_011526205.2 | XP_011524507.1 | 8 - 9 | 7535 | 147 | |
XM_011526206.2 | XP_011524508.1 | 8 - 9 | 4300 | 147 | |
XM_011526209.1 | XP_011524511.1 | 9 - 10 | 929 | 147 | |
XM_011526210.1 | XP_011524512.1 | 9 - 10 | 986 | 147 | |
XM_011526212.1 | XP_011524514.1 | 8 - 9 | 890 | 147 | |
XM_011526213.1 | XP_011524515.1 | 8 - 9 | 983 | 147 | |
XM_017026012.1 | XP_016881501.1 | 8 - 9 | 4147 | 147 | |
XM_017026013.1 | XP_016881502.1 | 9 - 10 | 4509 | 147 | |
XM_017026014.1 | XP_016881503.1 | 9 - 10 | 946 | 147 | |
GenBank mRNA | AA447511.1 | - | 2 - 3 | 147 | |
AK131454.1 | - | 8 - 9 | 737 | 147 |