The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017590.5 | NP_060060.3 | 11 - 12 | 1478 | 107 |
XM_011530016.2 | XP_011528318.1 | 11 - 12 | 1459 | 107 | |
GenBank mRNA | AB028954.1 | - | 8 - 9 | 1091 | 107 |
AF188530.1 | - | 11 - 12 | 1306 | 107 | |
AK315009.1 | - | 11 - 12 | 1420 | 107 | |
BC024313.2 | - | 2 - 3 | 145 | 107 | |
BC098378.1 | - | 11 - 12 | 1416 | 107 | |
BC152558.1 | - | 11 - 12 | 1456 | 107 |