The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017590.5 | NP_060060.3 | 2 - 3 | 333 | 99 |
XM_011530016.2 | XP_011528318.1 | 2 - 3 | 314 | 99 | |
GenBank mRNA | AK023849.1 | - | 2 - 3 | 121 | 99 |
AK225891.1 | - | 2 - 3 | 306 | 99 | |
AK315009.1 | - | 2 - 3 | 275 | 99 | |
BC098378.1 | - | 2 - 3 | 271 | 99 | |
BC152558.1 | - | 2 - 3 | 311 | 99 |