The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003770.4 | NP_003761.3 | 1 - 2 | 546 | 85 |
GenBank mRNA | AJ786655.1 | - | 1 - 2 | 382 | 85 |