The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010870.2 | NP_001010870.1 | 2 - 3 | 6417 | 140 |
GenBank mRNA | BX648686.1 | - | 2 - 3 | 2949 | 140 |
CR936792.1 | - | 2 - 3 | 4768 | 140 | |
EF185284.1 | - | 1 - 2 | 4548 | 140 |