Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4K14
Entrez Gene ID
122740
Gene Name
olfactory receptor family 4 subfamily K member 14
Gene Aliases
OR14-18, OR14-22
Chromosome Location
Chr.14: 20014261 - 20015193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004712.1 NP_001004712.1 1 - 1 344 124

View Full Product Details