The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170720.1 | NP_001164191.1 | 3 - 4 | 463 | 156 |
GenBank mRNA | AK302617.1 | - | 3 - 4 | 463 | 156 |